CAS RN 1620576-64-8
CAS RN 1620576-64-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express AZD-8835, MedChemExpress
MedChemExpress AZD8835 is a potent and selective inhibitor of PI3Kα and PI3Kδ with IC50s of 6.2 and 5.7 nM, respectively.
