CAS RN 1621-56-3
CAS RN 1621-56-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express 7-Methoxyisoflavone, MedChemExpress
MedChemExpress 7-Methoxyisoflavone is an isoflavone derivative and also an activator of adenosine monophosphate-activated protein kinase (AMPK).
