accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1640282-31-0

CH3ONOONH3COH3COH3C

CAS RN 1640282-31-0

IUPAC Name: 1-[7-(3,4-dimethoxyphenyl)-9-[(1-methylpiperidin-3-yl)methoxy]-2,3,4,5-tetrahydro-1,4-benzoxazepin-4-yl]propan-1-one
Synonyms: 1-[7-(3,4-dimethoxyphenyl)-9-[(1-methylpiperidin-3-yl)methoxy]-3,5-dihydro-2H-1,4-benzoxazepin-4-yl]propan-1-one
Molecular Weight (g/mol): 468.59
Molecular Formula: C27H36N2O5
InChi Key: YKNAKDFZAWQEEO-UHFFFAOYNA-N
SMILES: CCC(=O)N1CCOC2=C(C1)C=C(C=C2OCC1CCCN(C)C1)C1=CC(OC)=C(OC)C=C1
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products