accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 16603-18-2

H3COCH3OOH3COH3C

CAS RN 16603-18-2

IUPAC Name: 1-(3,4,5-trimethoxyphenyl)propan-2-one
Synonyms: 1-(3,4,5-trimethoxyphenyl)propan-2-one
Molecular Weight (g/mol): 224.26
Molecular Formula: C12H16O4
InChi Key: WDNDMVGQGRDIBC-UHFFFAOYSA-N
SMILES: COC1=CC(CC(C)=O)=CC(OC)=C1OC

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products