accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 167144-80-1

CH3NOOH3COHHCl

CAS RN 167144-80-1

IUPAC Name: 5-(2-{2-[2-(3-methoxyphenyl)ethyl]phenoxy}ethyl)-1-methylpyrrolidin-3-ol hydrochloride
Synonyms: 5-(2-{2-[2-(3-methoxyphenyl)ethyl]phenoxy}ethyl)-1-methylpyrrolidin-3-ol hydrochloride
Molecular Weight (g/mol): 391.94
Molecular Formula: C22H30ClNO3
InChi Key: OUKRSTJUAQEATQ-UHFFFAOYNA-N
SMILES: Cl.COC1=CC=CC(CCC2=CC=CC=C2OCCC2CC(O)CN2C)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products