CAS RN 168626-94-6
CAS RN 168626-94-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ Conivaptan Hydrochloride

Very high affinity vasopressin V1A and V2 antagonist; orally bioavailable
2
Medchem Express Conivaptan hydrochloride, MedChemExpress
MedChemExpress Conivaptan (hydrochloride) is a non-peptide antagonist of vasopressin receptor, with Ki values of 0.48 and 3.04 nM for rat liver V1A receptor and rat kidney V2 receptor respectively.
