accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 172531-37-2

OOOOOHNNN

CAS RN 172531-37-2

IUPAC Name: 2-{2-[2-(2-azidoethoxy)ethoxy]ethoxy}acetic acid
Synonyms: {2-[2-(2-azidoethoxy)ethoxy]ethoxy}acetic acid
Molecular Weight (g/mol): 233.22
Molecular Formula: C8H15N3O5
InChi Key: GIXBCECBLAEYKA-UHFFFAOYSA-N
SMILES: OC(=O)COCCOCCOCCN=[N+]=[N-]

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products