accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 17360-35-9

OHOOHOOHOOHOOHOOHOONNONN(E)(E)(E)

CAS RN 17360-35-9

IUPAC Name: tris((2E)-but-2-enedioic acid); bis(1-[4-(pyrrolidin-1-yl)but-2-yn-1-yl]pyrrolidin-2-one)
Synonyms: tris(fumaric acid); bis(oxotremorine)
Molecular Weight (g/mol): 760.79
Molecular Formula: C36H48N4O14
InChi Key: WLYYOFJEBGHKEC-VQYXCCSOSA-N
SMILES: OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.O=C1CCCN1CC#CCN1CCCC1.O=C1CCCN1CC#CCN1CCCC1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products