CAS RN 17990-42-0
CAS RN 17990-42-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Oleanonic acid, MedChemExpress
MedChemExpress Oleanonic acid (3-Oxooleanolic acid) is a triterpenoid, inhibits infection by HIV-1 in in vitro infected PBMC, naturally infected PBMC and monocyte/macrophages with EC50 of 22.7 mM, 24.6 mM and 57.4 mM, respectively.
