CAS RN 1813527-81-9
CAS RN 1813527-81-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express CCG215022, MedChemExpress
MedChemExpress CCG215022 is a G protein-coupled receptor kinases (GRKs) inhibitor with IC50s of 0.15±0.07 μM, 0.38±0.06 μM and 3.9±1 μM for GRK2, GRK5 and GRK1, respectively.
