CAS RN 18536-91-9
CAS RN 18536-91-9
IUPAC Name:
dodecyltriethoxysilane
Synonyms:
dodecyltriethoxysilane
Molecular Weight (g/mol):
332.6
Molecular Formula:
C18H40O3Si
InChi Key:
YGUFXEJWPRRAEK-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC[Si](OCC)(OCC)OCC
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar n-Dodecyltriethoxysilane, 95%
CAS: 18536-91-9 Molecular Formula: C18H40O3Si Molecular Weight (g/mol): 332.6 MDL Number: MFCD00048437 InChI Key: YGUFXEJWPRRAEK-UHFFFAOYSA-N Synonym: silane, dodecyltriethoxy,n-dodecyltriethoxysilane,lauryl triethoxy silane,dodecyl triethoxy silane,acmc-209emw,silane,dodecyltriethoxy,dodecyltriethoxysilane, technical PubChem CID: 87696 IUPAC Name: dodecyl(triethoxy)silane SMILES: CCCCCCCCCCCC[Si](OCC)(OCC)OCC