CAS RN 186205-33-4
CAS RN 186205-33-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress Cy2-SE (iodine)
MedChemExpress CY2-SE (Cyanine2 Succinimidyl Ester) is a dye for the labeling of amino-groups in peptides, proteins, and oligonucleotides.
