accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 18680-27-8

H3COHCH3OHH3C

CAS RN 18680-27-8

IUPAC Name: 2,6,6-trimethylbicyclo[3.1.1]heptane-2,3-diol
Synonyms: pinanediol
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: MOILFCKRQFQVFS-UHFFFAOYNA-N
SMILES: CC1(C)C2CC1C(C)(O)C(O)C2
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products