CAS RN 189188-57-6
CAS RN 189188-57-6
IUPAC Name:
(2E)-but-2-enedioic acid; N-({[(3Z)-5-(hydroxymethyl)-3H-indol-3-ylidene]methyl}amino)-N''-pentylguanidine
Synonyms:
tegaserod
Molecular Weight (g/mol):
417.47
Molecular Formula:
C20H27N5O5
InChi Key:
PINGCSUEYQYSEB-GFQXDJOGSA-N
SMILES:
OC(=O)\C=C\C(O)=O.CCCCCN=C(N)NN\C=C1/C=NC2=CC=C(CO)C=C12
1
–
1
of
1
results
1
Medchem Express Tegaserod maleate, MedChemExpress
MedChemExpress Tegaserod maleate is a selective 5-HT4 receptor partial agonist and a 5-HT2B receptor antagonist. Tegaserod maleate exhibits a promotile effect throughout the gastrointestinal (GI) tract.
