accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 189281-66-1

CH3OONClClF

CAS RN 189281-66-1

IUPAC Name: methyl 2,6-dichloro-5-fluoropyridine-3-carboxylate
Synonyms: methyl 2,6-dichloro-5-fluoropyridine-3-carboxylate
Molecular Weight (g/mol): 224.01
Molecular Formula: C7H4Cl2FNO2
InChi Key: WADLLLSMEPLCNO-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(F)=C(Cl)N=C1Cl

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products