CAS RN 19083-00-2
CAS RN 19083-00-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MedChemExpress Gracillin
MedChemExpress Gracillin is a kind of steroidal saponin isolated from the root bark of wild yam Dioscorea nipponica with antitumor agent.

2
MedChemExpress Gracillin
MedChemExpress Gracillin is a kind of steroidal saponin isolated from the root bark of wild yam Dioscorea nipponica with antitumor agent.
