CAS RN 2288709-96-4
CAS RN 2288709-96-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express AZ32, MedChemExpress
MedChemExpress AZ32 is an orally bioavailable and blood-brain barrier-penetrating ATM inhibitor with an IC50 of IC50 of 0.31 μM for ATM in cell.
