CAS RN 2438-72-4
CAS RN 2438-72-4
IUPAC Name:
2-(4-butoxyphenyl)-N-hydroxyacetamide
Synonyms:
bufexamic acid
Molecular Weight (g/mol):
223.27
Molecular Formula:
C12H17NO3
InChi Key:
MXJWRABVEGLYDG-UHFFFAOYSA-N
SMILES:
CCCCOC1=CC=C(CC(=O)NO)C=C1
1
–
1
of
1
results
1
Medchem Express Bufexamac, MedChemExpress
MedChemExpress Bufexamac is a class IIB histone deacetylases (HDAC6 and HDAC10) inhibitor used as an anti-inflammatory agent.
