accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 2613-32-3

FFClNH2

CAS RN 2613-32-3

IUPAC Name: 2-chloro-4,5-difluoroaniline
Synonyms: 2-chloro-4,5-difluoroaniline
Molecular Weight (g/mol): 163.55
Molecular Formula: C6H4ClF2N
InChi Key: ZNDPHOPPLHZBMV-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(F)C=C1Cl

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products