CAS RN 2731-73-9
CAS RN 2731-73-9
IUPAC Name:
(2R)-2-amino-3-chloropropanoic acid
Synonyms:
3-chloro-L-alanine
Molecular Weight (g/mol):
123.54
Molecular Formula:
C3H6ClNO2
InChi Key:
ASBJGPTTYPEMLP-REOHCLBHSA-N
SMILES:
N[C@@H](CCl)C(O)=O
1
–
2
of
2
results
2
Medchem Express β-Chloro-L-alanine, MedChemExpress
MedChemExpress β-Chloro-L-alanine is a bacteriostatic amino acid analog which inhibits a number of enzymes, including threonine deaminase and alanine racemase.
