accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 274682-91-6

CH3OFF

CAS RN 274682-91-6

IUPAC Name: 1-(2,4-difluorophenyl)propan-2-one
Synonyms: 1-(2,4-difluorophenyl)propan-2-one
Molecular Weight (g/mol): 170.16
Molecular Formula: C9H8F2O
InChi Key: XVHHDTNIOCSTLH-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=C(F)C=C1F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products