CAS RN 292151-97-4
CAS RN 292151-97-4
IUPAC Name:
1-[3-methyl-5-(trifluoromethyl)phenyl]methanamine
Synonyms:
1-[3-methyl-5-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol):
189.18
Molecular Formula:
C9H10F3N
InChi Key:
ZESXWCTWKWVJGM-UHFFFAOYSA-N
SMILES:
CC1=CC(CN)=CC(=C1)C(F)(F)F
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 3-Methyl-5-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™
CAS: 292151-97-4 Molecular Formula: C9H10F3N Molecular Weight (g/mol): 189.181 MDL Number: MFCD06213553 InChI Key: ZESXWCTWKWVJGM-UHFFFAOYSA-N Synonym: 3-methyl-5-trifluoromethyl benzylamine,1-3-methyl-5-trifluoromethyl phenyl methanamine,3-methyl-5-trifluoromethyl phenyl methanamine,3-methyl-5-trifluoromethyl benzyl amine PubChem CID: 18546522 IUPAC Name: [3-methyl-5-(trifluoromethyl)phenyl]methanamine SMILES: CC1=CC(=CC(=C1)C(F)(F)F)CN