CAS RN 300816-15-3
CAS RN 300816-15-3
IUPAC Name:
6-methyl-1'-[2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethyl]-1,2-dihydrospiro[3,1-benzoxazine-4,4'-piperidin]-2-one
Synonyms:
6-methyl-1'-[2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethyl]-1H-spiro[3,1-benzoxazine-4,4'-piperidin]-2-one
Molecular Weight (g/mol):
417.51
Molecular Formula:
C25H27N3O3
InChi Key:
ODNICNWASXKNNQ-UHFFFAOYSA-N
SMILES:
CC1=C(CCN2CCC3(CC2)OC(=O)NC2=C3C=C(C)C=C2)N=C(O1)C1=CC=CC=C1
1
–
2
of
2
results
2
Medchem Express RS 504393, MedChemExpress
MedChemExpress RS 504393 is a selective CCR2 chemokine receptor antagonist (IC50 values are 89 nM and > 100 μM for inhibition of human recombinant CCR2 and CCR1 receptors respectively).
