accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 302912-19-2

NCOFFFF

CAS RN 302912-19-2

IUPAC Name: 1-fluoro-3-isocyanato-5-(trifluoromethyl)benzene
Synonyms: 1-fluoro-3-isocyanato-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 205.11
Molecular Formula: C8H3F4NO
InChi Key: WGSXKYXKAARAKD-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(=C1)N=C=O)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products