CAS RN 31083-55-3
CAS RN 31083-55-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express PRT4165, MedChemExpress
MedChemExpress PRT4165 is a potent inhibitor of PRC1-mediated H2A ubiquitylation.
