accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 31137-74-3

CH3(R)(S)NH3OOOO

CAS RN 31137-74-3

IUPAC Name: (2S,4R)-2-azaniumyl-4-methylpentanedioate
Synonyms: (2S,4R)-2-ammonio-4-methylpentanedioate
Molecular Weight (g/mol): 160.15
Molecular Formula: C6H10NO4
InChi Key: KRKRAOXTGDJWNI-DMTCNVIQSA-M
SMILES: C[C@H](C[C@H]([NH3+])C([O-])=O)C([O-])=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products