accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 3118-68-1

SOONH2N

CAS RN 3118-68-1

IUPAC Name: 3-cyanobenzene-1-sulfonamide
Synonyms: benzenesulfonamide, 3-cyano-
Molecular Weight (g/mol): 182.2
Molecular Formula: C7H6N2O2S
InChi Key: LTOFQPCBIZPXFH-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=CC(=C1)C#N

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products