CAS RN 313397-13-6
CAS RN 313397-13-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Reversan, MedChemExpress
MedChemExpress Reversan (CBLC4H10) is a potent and nontoxic multidrug resistance-associated protein 1 (MRP1) and P-glycoprotein (Pgp) inhibitor.
