accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 33421-40-8

NH2N

CAS RN 33421-40-8

IUPAC Name: 5-phenylpyridin-2-amine
Synonyms: 5-phenylpyridin-2-amine
Molecular Weight (g/mol): 170.22
Molecular Formula: C11H10N2
InChi Key: OAPVIBHQRYFYSE-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=N1)C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products