CAS RN 348-52-7
CAS RN 348-52-7
IUPAC Name:
1-fluoro-2-iodobenzene
Synonyms:
benzene, 1-fluoro-2-iodo-
Molecular Weight (g/mol):
222
Molecular Formula:
C6H4FI
InChi Key:
TYHUGKGZNOULKD-UHFFFAOYSA-N
SMILES:
FC1=CC=CC=C1I
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Alfa Aesar 1-Fluoro-2-iodobenzene, 99%, stab. with copper
Thermo Scientific Alfa Aesar 1-Fluoro-2-iodobenzene, 99%, stab. with copper
CAS: 348-52-7 Molecular Formula: C6H4FI Molecular Weight (g/mol): 222.00 MDL Number: MFCD00001031 InChI Key: TYHUGKGZNOULKD-UHFFFAOYSA-N Synonym: 2-fluoroiodobenzene,o-fluoroiodobenzene,o-iodofluorobenzene,benzene, 1-fluoro-2-iodo,2-iodofluorobenzene,3-fluoro-2-iodobenzene,1-fluoro-2-iodo-benzene,2-fluoro-iodobenzene,2-fluoro-1-iodobenzene PubChem CID: 67673 IUPAC Name: 1-fluoro-2-iodobenzene SMILES: FC1=CC=CC=C1I