accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 367954-99-2

CH3OOHFF

CAS RN 367954-99-2

IUPAC Name: 2,4-difluoro-5-methylbenzoic acid
Synonyms: 2,4-difluoro-5-methylbenzoic acid
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: YWQRTHMEMOUEPW-UHFFFAOYSA-N
SMILES: CC1=C(F)C=C(F)C(=C1)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products