CAS RN 3730-60-7
CAS RN 3730-60-7
IUPAC Name:
2,5-dimethylhexan-2-ol
Synonyms:
2-hexanol, 2,5-dimethyl-, (S)-
Molecular Weight (g/mol):
130.23
Molecular Formula:
C8H18O
InChi Key:
JPUIYNHIEXIFMV-UHFFFAOYSA-N
SMILES:
CC(C)CCC(C)(C)O
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 2,5-Dimethyl-2-hexanol, 98%
CAS: 3730-60-7 Molecular Formula: C8H18O Molecular Weight (g/mol): 130.23 MDL Number: MFCD00021814 InChI Key: JPUIYNHIEXIFMV-UHFFFAOYSA-N Synonym: 2,5-dimethyl-2-hexanol,2-hexanol, 2,5-dimethyl-, s,2-hexanol, 2,5-dimethyl,2-hexanol, 2,5-dimethyl-, s-+,acmc-209is7,2-hexanol,2,5-dimethyl,2-hexanol,5-dimethyl-, s,2-hexanol,5-dimethyl-, s-+ PubChem CID: 19506 IUPAC Name: 2,5-dimethylhexan-2-ol SMILES: CC(C)CCC(C)(C)O