CAS RN 41748-71-4
CAS RN 41748-71-4
IUPAC Name:
1H-indazol-4-amine
Synonyms:
1H-indazol-4-amine
Molecular Weight (g/mol):
133.15
Molecular Formula:
C7H7N3
InChi Key:
MDELYEBAXHZXLZ-UHFFFAOYSA-N
SMILES:
NC1=C2C=NNC2=CC=C1