accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 50-43-1

OOClClCl

CAS RN 50-43-1

IUPAC Name: 2,4,6-trichlorobenzoate
Synonyms: 2,4,6-trichlorobenzoate
Molecular Weight (g/mol): 224.44
Molecular Formula: C7H2Cl3O2
InChi Key: RAFFVQBMVYYTQS-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=C(Cl)C=C(Cl)C=C1Cl

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products