accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 5424-47-5

HClH3CNCH3OS

CAS RN 5424-47-5

IUPAC Name: hydrogen 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-one chloride
Synonyms: hydrogen 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-one chloride
Molecular Weight (g/mol): 219.73
Molecular Formula: C9H14ClNOS
InChi Key: HPVHJPMLORARSR-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCC(=O)C1=CC=CS1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products