accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 627-00-9

HOOCl

CAS RN 627-00-9

IUPAC Name: 4-chlorobutanoic acid
Synonyms: butanoic acid, 4-chloro-
Molecular Weight (g/mol): 122.55
Molecular Formula: C4H7ClO2
InChi Key: IPLKGJHGWCVSOG-UHFFFAOYSA-N
SMILES: OC(=O)CCCCl

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products