CAS RN 707-60-8
CAS RN 707-60-8
IUPAC Name:
4-bromo-N,N-dimethylbenzene-1-sulfonamide
Synonyms:
4-bromo-N,N-dimethylbenzenesulfonamide
Molecular Weight (g/mol):
264.14
Molecular Formula:
C8H10BrNO2S
InChi Key:
NQAUNPZZVCXYEJ-UHFFFAOYSA-N
SMILES:
CN(C)S(=O)(=O)C1=CC=C(Br)C=C1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 4-Bromo-N,N-dimethylbenzenesulfonamide, 97%, Thermo Scientific™
CAS: 707-60-8 Molecular Formula: C8H10BrNO2S Molecular Weight (g/mol): 264.137 MDL Number: MFCD00017834 InChI Key: NQAUNPZZVCXYEJ-UHFFFAOYSA-N Synonym: 4-bromo-n,n-dimethyl-benzenesulfonamide,benzenesulfonamide, 4-bromo-n,n-dimethyl,p-bromo-n,n-dimethylbenzenesulfonamide,4-bromobenzenedimethylsulfonamide,4-bromo-n,n-dimethylbenzene-1-sulfonamide,n,n-dimethyl 4-bromobenzenesulfonamide,benzenesulfonamide, p-bromo-n,n-dimethyl,n,n-dimethyl-4-bromobenzenesulfonamide,4-bromo-n,n-dimethylbenzenesulphonamide,acmc-209og0 PubChem CID: 12818 IUPAC Name: 4-bromo-N,N-dimethylbenzenesulfonamide SMILES: CN(C)S(=O)(=O)C1=CC=C(C=C1)Br