accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 71022-43-0

OHNOONOO

CAS RN 71022-43-0

IUPAC Name: (3,5-dinitrophenyl)methanol
Synonyms: (3,5-dinitrophenyl)methanol
Molecular Weight (g/mol): 198.13
Molecular Formula: C7H6N2O5
InChi Key: GPHYIQCSMDYRGJ-UHFFFAOYSA-N
SMILES: OCC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products