accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 87394-64-7

NNHNONH2

CAS RN 87394-64-7

IUPAC Name: 2-(piperazin-1-yl)pyridine-3-carboxamide
Synonyms: 2-(piperazin-1-yl)pyridine-3-carboxamide
Molecular Weight (g/mol): 206.25
Molecular Formula: C10H14N4O
InChi Key: RYNARQDUUQTTFJ-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(N=CC=C1)N1CCNCC1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products