accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 90292-67-4

FFBrCl

CAS RN 90292-67-4

IUPAC Name: 2-(bromomethyl)-3-chloro-1,4-difluorobenzene
Synonyms: 2-(bromomethyl)-3-chloro-1,4-difluorobenzene
Molecular Weight (g/mol): 241.46
Molecular Formula: C7H4BrClF2
InChi Key: KAADKOQBKOGBAA-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(CBr)=C1Cl

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products