accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 97-50-7

H3COOCH3ClH2N

CAS RN 97-50-7

IUPAC Name: 5-chloro-2,4-dimethoxyaniline
Synonyms: aniline,2,4-dimethoxy-5-chloro
Molecular Weight (g/mol): 187.62
Molecular Formula: C8H10ClNO2
InChi Key: OLCMNCWEUMBNIS-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(Cl)C=C1N

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products