CAS RN 117-83-9
IUPAC Namn:
1,2-bis(2-butoxyethyl) benzene-1,2-dicarboxylate
Synonymer:
palatinol dibutylglycol phthalate palatinol K di(butoxyethyl)phthalate phthalic acid dibutoxy ethyl ester kronisol kesscoflex butyl "cellosolve" phthalate dibutyl cellosolve phthalate bis(2-butoxyethyl)phthalate
Molekylvikt (g/mol):
366.45
Molekylär sammansättning:
C20H30O6
InChi Key:
CMCJNODIWQEOAI-UHFFFAOYSA-N
SMILES:
CCCCOCCOC(=O)C1=CC=CC=C1C(=O)OCCOCCCC
1
–
2
av
2
Resultat
1
Bis(2-butoxyethyl) Phthalate (~85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Bis(2-butoxyethyl) Phthalate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.