CAS RN 1246343-36-1
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
4
av
4
Resultat
1
Praziquantel-d11, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Praziquantel-d11, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Praziquantel D11, MedChemExpress
MedChemExpress Praziquantel D11 is the deuterium labeled Praziquantel, which is an anthelmintic.