CAS RN 1391053-55-6
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
4
av
4
Resultat
1
3’-[4-(5-Chloro-2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)-1-piperidinyl]propyl Domperidine (Domperidine Impurity F), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3’-[4-(5-Chloro-2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)-1-piperidinyl]propyl Domperidine (Domperidine Impurity F), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3’-[4-(5-Chloro-2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)-1-piperidinyl]propyl Domperidine (Domperidine Impurity F), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
1,3-Bis[3-[4-(5-chloro-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)piperidin-1-yl]propyl]-1,3-dihydro-2H-benzimidazol-2-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.