CAS RN 1440209-96-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
Tocris Bioscience™ BRD 73954
Dual histone deacetylase (HDAC) 6/8 inhibitor
2
BRD73954, MedChemExpress
MedChemExpress BRD73954 ia a potent and selective HDAC inhibitor with IC50 of 36 nM and 120 nM for HDAC6 and HDAC8, respectively.