CAS RN 14816-18-3
IUPAC Namn:
(Z)-benzenecarbonimidoyl cyanide
Synonymer:
phoxim (iso)
Molekylvikt (g/mol):
298.3
Molekylär sammansättning:
C12H15N2O3PS
InChi Key:
ATROHALUCMTWTB-WYMLVPIESA-N
SMILES:
CCOP(=S)(OCC)O\N=C(/C#N)C1=CC=CC=C1
1
–
5
av
5
Resultat
1
Phoxim, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Phoxim 1000 μg/mL in Acetone, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
3
Phoxim, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Phoxim, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.