CAS RN 1686-14-2
IUPAC Namn:
2,7,7-trimethyl-3-oxatricyclo[4.1.1.0²,⁴]octane
Synonymer:
α-pinene oxide 2-pinene oxide α-pinene epoxide
Molekylvikt (g/mol):
152.24
Molekylär sammansättning:
C10H16O
InChi Key:
NQFUSWIGRKFAHK-UHFFFAOYNA-N
SMILES:
CC12OC1CC1CC2C1(C)C
1
–
3
av
3
Resultat
1
a-Pinene Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Alpha-Pinene Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Alpha-Pinene Oxide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.