CAS RN 17146-86-0
IUPAC Namn:
(carboxymethyl)trimethylazanium hydroxide
Synonymer:
(carboxymethyl)trimethylammonium hydroxide inner salt
Molekylvikt (g/mol):
135.16
Molekylär sammansättning:
C5H13NO3
InChi Key:
NJZRLXNBGZBREL-UHFFFAOYSA-N
SMILES:
[OH-].C[N+](C)(C)CC(O)=O