CAS RN 1906919-67-2
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
BAY-598, MedChemExpress
MedChemExpress BAY-598 is selective small molecule inhibitor of SMYD2 with an IC50 of 27 nM.
2
Tocris Bioscience™ BAY 598
Potent and selective SMYD2 inhibitor