CAS RN 1912-31-8
IUPAC Namn:
propyl methanesulfonate
Synonymer:
propyl methanesulfonate
Molekylvikt (g/mol):
138.18
Molekylär sammansättning:
C4H10O3S
InChi Key:
DKORSYDQYFVQNS-UHFFFAOYSA-N
SMILES:
CCCOS(C)(=O)=O
1
–
3
av
3
Resultat
1
Propyl Methanesulfonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Propyl Methanesulfonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Propyl Methanesulfonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.